Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144070.1 | NP_001137542.1 | 2 | 262 | ||
NM_003786.3 | NP_003777.2 | 2 | 262 | |||
XM_005257763.3 | XP_005257820.1 | 2 | 276 | |||
XM_011525422.2 | XP_011523724.1 | 2 | 276 | |||
XM_011525423.1 | XP_011523725.1 | 2 | 276 | |||
XM_011525425.2 | XP_011523727.1 | 1 | 39 | |||
XM_017025266.1 | XP_016880755.1 | 2 | 276 | |||
GenBank mRNA | AB010887.1 | - | 2 | 196 | ||
AB208954.1 | - | 2 | 202 | |||
AF009670.1 | - | 2 | 252 | |||
AF083552.1 | - | 2 | 199 | |||
AF085690.1 | - | 2 | 218 | |||
AF085691.1 | - | 2 | 218 | |||
AF085692.1 | - | 2 | 218 | |||
AF104943.1 | - | 2 | 186 | |||
BC046126.1 | - | 2 | 262 | |||
BC050370.1 | - | 2 | 187 | |||
BC058907.1 | - | 2 | 221 | |||
BC104952.1 | - | 2 | 214 | |||
BC137347.1 | - | 2 | 213 | 9021345 | ||
BC137348.1 | - | 2 | 213 | 9021346 | ||
Y17151.2 | - | 2 | 224 |