Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003818.3 | NP_003809.1 | 13 | 2731 | ||
XM_006723660.1 | XP_006723723.1 | 13 | 2742 | |||
XM_011529390.1 | XP_011527692.1 | 14 | 3054 | |||
GenBank mRNA | AF069532.1 | - | 12 | 2327 | ||
AK027695.1 | - | 13 | 2439 | |||
BC025751.1 | - | 14 | 2053 | 5199748 | ||
Y16521.1 | - | 13 | 2654 |