Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004077.2 | NP_004068.2 | 4 | 418 | ||
GenBank mRNA | AF053631.1 | - | 4 | 393 | ||
AF147410.1 | - | 2 | 118 | |||
AK074956.1 | - | 4 | 266 | |||
AK095856.1 | - | 4 | 253 | |||
AK293263.1 | - | 2 | 203 | |||
AK296249.1 | - | 5 | 398 | |||
BC000105.1 | - | 2 | 58 | |||
BC010106.1 | - | 4 | 346 | 3542491 | ||
BC072016.1 | - | 4 | 261 | 6484456 | ||
BI258893.1 | - | 5 | 445 | |||
BX640838.1 | - | 5 | 475 | |||
CB990178.1 | - | 4 | 445 |