Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004170.5 | NP_004161.4 | 7 | 1001 | ||
XM_011518007.1 | XP_011516309.1 | 8 | 1021 | |||
XM_011518008.2 | XP_011516310.1 | 8 | 8266 | |||
XM_011518009.2 | XP_011516311.1 | 7 | 8196 | |||
XM_011518010.1 | XP_011516312.1 | 6 | 811 | |||
XM_017015042.1 | XP_016870531.1 | 8 | 1021 | |||
XM_017015043.1 | XP_016870532.1 | 7 | 962 | |||
GenBank mRNA | AB008536.1 | - | 7 | 939 | ||
AK313013.1 | - | 7 | 902 | |||
BC033040.1 | - | 7 | 987 | 5261168 | ||
U03506.1 | - | 7 | 766 | |||
U06469.1 | - | 7 | 787 | |||
U08989.1 | - | 7 | 913 |