Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281501.1 | NP_001268430.1 | 12 | 2952 | ||
NM_001281502.1 | NP_001268431.1 | 12 | 3021 | |||
NM_004265.3 | NP_004256.1 | 12 | 3293 | |||
GenBank mRNA | AF084559.1 | - | 12 | 3085 | ||
AF108658.1 | - | 12 | 2878 | |||
AF126799.1 | - | 12 | 2952 | |||
AK027459.1 | - | 6 | 2088 | |||
AK027513.1 | - | 6 | 2107 | |||
AK027577.1 | - | 6 | 2132 | |||
AK074925.1 | - | 10 | 2500 | |||
AK074939.1 | - | 12 | 3025 | |||
AK074991.1 | - | 12 | 2761 | |||
AK123240.1 | - | 7 | 2922 | |||
AL050118.1 | - | 10 | 2537 |