Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004273.4 | NP_004264.2 | 3 | 4867 | ||
XM_006718075.3 | XP_006718138.1 | 3 | 5100 | |||
XM_011540369.2 | XP_011538671.1 | 3 | 6793 | |||
GenBank mRNA | AB017915.1 | - | 3 | 4865 | ||
AK025251.1 | - | 1 | 203 |