Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004582.3 | NP_004573.2 | 9 | 1078 | ||
NR_073562.1 | 11 | 1273 | ||||
GenBank mRNA | AK127197.1 | - | 3 | 1930 | ||
AK128861.1 | - | 7 | 3681 | |||
AK293057.1 | - | 11 | 1273 | |||
BC020790.1 | - | 9 | 1071 | 4290610 | ||
U49245.1 | - | 8 | 1000 | |||
X98001.1 | - | 8 | 1024 | |||
Y08201.1 | - | 8 | 1003 |