Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271765.1 | NP_001258694.1 | 7 | 1955 | ||
NM_004695.3 | NP_004686.1 | 7 | 1889 | |||
XM_005257790.1 | XP_005257847.1 | 6 | 1831 | |||
XM_011525462.1 | XP_011523764.1 | 6 | 1779 | |||
XM_017025293.1 | XP_016880782.1 | 6 | 3616 | |||
GenBank mRNA | AK000416.1 | - | 7 | 1955 | ||
AK092512.1 | - | 6 | 1998 | |||
BC009684.1 | - | 7 | 1870 | 3895881 | ||
BC033611.1 | - | 6 | 1779 | 5218712 | ||
U59299.1 | - | 5 | 1643 |