Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC33A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190992.1 | NP_001177921.1 | 1 | 959 | ||
NM_004733.3 | NP_004724.1 | 1 | 959 | |||
XM_011513311.2 | XP_011511613.1 | 1 | 929 | |||
XM_017007462.1 | XP_016862951.1 | 1 | 929 | |||
XM_017007463.1 | XP_016862952.1 | 1 | 927 | |||
XM_017007464.1 | XP_016862953.1 | 1 | 916 | |||
GenBank mRNA | AK312268.1 | - | 1 | 929 | ||
BC014416.1 | - | 1 | 906 | 4559958 | ||
D88152.1 | - | 1 | 884 |