Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037277.1 | NP_001032354.1 | 4 | 334 | ||
NR_036605.1 | 3 | 405 | ||||
GenBank mRNA | AB016043.1 | - | 4 | 419 | ||
AB017971.1 | - | 3 | 250 | |||
AB019036.1 | - | 4 | 460 | |||
AF057698.1 | - | 4 | 488 | |||
AF125394.1 | - | 4 | 397 | |||
AK025139.1 | - | 4 | 482 | |||
AK293278.1 | - | 3 | 388 | |||
BC005252.1 | - | 4 | 482 | 4051481 | ||
BC067768.1 | - | 4 | 422 | 5295932 | ||
BI462253.1 | - | 4 | 509 | |||
BI520491.1 | - | 4 | 337 | |||
BI963107.1 | - | 3 | 355 | |||
CB216084.1 | - | 4 | 500 |