Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101662.1 | NP_001095132.1 | 2 | 699 | ||
NM_001242361.1 | NP_001229290.1 | 2 | 94 | |||
NM_002525.2 | NP_002516.2 | 2 | 699 | |||
XM_011541522.1 | XP_011539824.1 | 3 | 688 | |||
XM_011541523.1 | XP_011539825.1 | 2 | 263 | |||
XM_011541525.2 | XP_011539827.1 | 2 | 126 | |||
XM_017001375.1 | XP_016856864.1 | 2 | 123 | |||
GenBank mRNA | AK299271.1 | - | 2 | 149 | ||
AK302616.1 | - | 2 | 123 | |||
AK311631.1 | - | 2 | 96 | |||
AK312318.1 | - | 2 | 511 | |||
AY049784.1 | - | 2 | 417 | |||
AY360265.1 | - | 2 | 73 | |||
BC008775.2 | - | 2 | 476 | 3138712 | ||
DA245717.1 | - | 2 | 573 | |||
DB084774.1 | - | 2 | 95 | |||
U64898.1 | - | 2 | 474 | |||
X93207.1 | - | 2 | 512 | |||
X93209.1 | - | 2 | 512 |