Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004888.3 | NP_004879.1 | 3 | 547 | ||
GenBank mRNA | AF038954.1 | - | 3 | 486 | ||
BC008452.1 | - | 3 | 473 | 4246202 | ||
BI596632.1 | - | 3 | 524 |