Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078175.2 | NP_001071643.1 | 14 | 2092 | ||
NM_001078177.1 | NP_001071645.1 | 14 | 2287 | |||
NM_001304462.1 | NP_001291391.1 | 14 | 2225 | |||
NM_001304463.1 | NP_001291392.1 | 13 | 1985 | |||
NM_001304465.1 | NP_001291394.1 | 13 | 2128 | |||
NM_001304466.1 | NP_001291395.1 | 13 | 2125 | |||
XM_005248876.4 | XP_005248933.1 | 13 | 1950 | |||
XM_005248878.3 | XP_005248935.1 | 14 | 2122 | |||
XM_005248879.3 | XP_005248936.1 | 14 | 2056 | |||
XM_005248880.3 | XP_005248937.1 | 14 | 2033 | |||
XM_005248881.3 | XP_005248938.1 | 13 | 2130 | |||
XM_005248882.3 | XP_005248939.1 | 13 | 2073 | |||
XM_011514341.2 | XP_011512643.1 | 14 | 2146 | |||
GenBank mRNA | AK090491.1 | - | 14 | 2094 | ||
AK090615.1 | - | 14 | 2113 | |||
AX775997.1 | - | 13 | 1974 | |||
BC001382.2 | - | 13 | 1898 | 3051441 | ||
BC008954.2 | - | 14 | 1971 | 3010092 | ||
U81375.1 | - | 13 | 1974 |