Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145961.1 | NP_001139433.1 | 3 | 383 | ||
NM_001145962.1 | NP_001139434.1 | 2 | 288 | |||
NM_001145963.1 | NP_001139435.1 | 3 | 262 | |||
NM_001145964.1 | NP_001139436.1 | 3 | 253 | |||
NM_005072.4 | NP_005063.1 | 3 | 383 | |||
GenBank mRNA | AF047338.1 | - | 3 | 242 | ||
AF054505.1 | - | 3 | 242 | |||
AF054506.1 | - | 3 | 242 | |||
AK097808.1 | - | 4 | 388 | |||
AK293906.1 | - | 3 | 292 | |||
AK293956.1 | - | 2 | 178 | |||
AK299042.1 | - | 3 | 169 | |||
AK302790.1 | - | 3 | 262 | |||
AK311073.1 | - | 3 | 283 | |||
AK316415.1 | - | 3 | 243 | |||
BC021193.2 | - | 3 | 311 | 3349710 | ||
BC035480.1 | - | 3 | 1564 | |||
DC321966.1 | - | 3 | 384 | |||
DC333904.1 | - | 3 | 253 | |||
DC402650.1 | - | 3 | 262 | |||
U55054.1 | - | 3 | 297 |