Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005094.3 | NP_005085.2 | 6 | 1130 | ||
XM_017014222.1 | XP_016869711.1 | 7 | 1003 | |||
GenBank mRNA | AF055899.1 | - | 5 | 845 | ||
AK027504.1 | - | 5 | 710 | |||
AK074831.1 | - | 3 | 178 | |||
AK290222.1 | - | 6 | 1130 | |||
AK314681.1 | - | 6 | 970 | |||
BC065003.1 | - | 6 | 1129 | 6023438 |