Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005110.3 | NP_005101.1 | 19 | 2575 | ||
XM_006714942.2 | XP_006715005.1 | 18 | 2362 | |||
XM_017010100.1 | XP_016865589.1 | 18 | 2460 | |||
GenBank mRNA | AB016789.1 | - | 19 | 2504 | ||
AK001242.1 | - | 9 | 1457 | |||
AK022507.1 | - | 19 | 2515 | |||
BC000012.2 | - | 19 | 2513 | 3506723 |