Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005135.2 | NP_005126.1 | 1 | 219 | ||
GenBank mRNA | AF108831.1 | - | 1 | 128 | ||
AK315283.1 | - | 1 | 191 | |||
BC126241.1 | - | 1 | 127 | 8991957 | ||
BC126243.1 | - | 1 | 127 | 8991959 |