Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002533.3 | NP_002524.2 | 6 | 542 | ||
NM_206840.2 | NP_996671.1 | 5 | 468 | |||
XM_011544196.1 | XP_011542498.1 | 6 | 525 | |||
XM_011544198.1 | XP_011542500.1 | 5 | 480 | |||
XM_011544199.1 | XP_011542501.1 | 5 | 434 | |||
XM_011544200.1 | XP_011542502.1 | 4 | 408 | |||
XM_017001378.1 | XP_016856867.1 | 7 | 634 | |||
XM_017001379.1 | XP_016856868.1 | 7 | 634 | |||
XM_017001380.1 | XP_016856869.1 | 7 | 634 | |||
XM_017001381.1 | XP_016856870.1 | 7 | 631 | |||
XM_017001382.1 | XP_016856871.1 | 6 | 564 | |||
XM_017001383.1 | XP_016856872.1 | 6 | 564 | |||
XM_017001384.1 | XP_016856873.1 | 7 | 634 | |||
XM_017001385.1 | XP_016856874.1 | 7 | 634 | |||
XM_017001386.1 | XP_016856875.1 | 4 | 408 | |||
GenBank mRNA | AK297070.1 | - | 6 | 501 | ||
AK298826.1 | - | 5 | 420 | |||
AW997091.1 | - | 3 | 287 | |||
BC005897.1 | - | 6 | 501 | |||
BP284587.1 | - | 6 | 542 | |||
BQ883523.1 | - | 1 | 62 | |||
U68140.1 | - | 6 | 536 | |||
U78772.1 | - | 5 | 449 |