Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005328.2 | NP_005319.1 | 2 | 628 | ||
GenBank mRNA | AK297724.1 | - | 2 | 674 | ||
BC069353.1 | - | 2 | 162 | 7262337 | ||
BC109071.1 | - | 2 | 533 | 40034202 | ||
BC109072.1 | - | 2 | 533 | 40034203 | ||
U54804.1 | - | 2 | 625 |