Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005385.3 | NP_005376.2 | 5 | 408 | ||
XM_005265173.1 | XP_005265230.1 | 5 | 403 | |||
XM_006713171.1 | XP_006713234.1 | 5 | 403 | |||
XM_006713173.2 | XP_006713236.1 | 5 | 397 | |||
XM_011533747.2 | XP_011532049.1 | 5 | 363 | |||
XM_011533750.2 | XP_011532052.1 | 5 | 363 | |||
XM_017006474.1 | XP_016861963.1 | 5 | 403 | |||
XM_017006475.1 | XP_016861964.1 | 5 | 379 | |||
XM_017006480.1 | XP_016861969.1 | 5 | 363 | |||
GenBank mRNA | AB209888.1 | - | 4 | 897 | ||
AK292017.1 | - | 5 | 381 | |||
BC072680.1 | - | 5 | 351 | |||
BC144518.1 | - | 5 | 381 | |||
BX538094.1 | - | 945 | ||||
BX957218.1 | - | 5 | 378 | |||
CR749571.1 | - | 5 | 378 | |||
L04288.2 | - | 5 | 449 |