Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005415.4 | NP_005406.3 | 2 | 731 | ||
XM_017004768.1 | XP_016860257.1 | 1 | 444 | |||
XM_017004769.1 | XP_016860258.1 | 2 | 731 | |||
XM_017004770.1 | XP_016860259.1 | 2 | 702 | |||
XM_017004771.1 | XP_016860260.1 | 2 | 702 | |||
GenBank mRNA | AK312738.1 | - | 2 | 650 | ||
BC019944.1 | - | 2 | 638 | 3918690 | ||
L20859.1 | - | 2 | 562 |