Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005482.2 | NP_005473.1 | 4 | 348 | ||
GenBank mRNA | AF022913.1 | - | 4 | 320 | ||
AK313015.1 | - | 4 | 331 | |||
BC020737.1 | - | 4 | 327 | 4722399 | ||
DA263434.1 | - | 4 | 348 | |||
Y07596.1 | - | 4 | 307 |