Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287189.1 | NP_001274118.1 | 4 | 622 | ||
NM_001287190.1 | NP_001274119.1 | 4 | 468 | |||
NM_002539.2 | NP_002530.1 | 4 | 730 | |||
GenBank mRNA | AK292352.1 | - | 4 | 603 | ||
AK312766.1 | - | 2 | 170 | |||
BC025296.1 | - | 4 | 470 | 5088190 | ||
BI463057.1 | - | 4 | 468 | |||
DQ185043.1 | - | 1 | 24 | |||
M16650.2 | - | 3 | 256 | |||
X55362.1 | - | 4 | 471 |