Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005502.3 | NP_005493.2 | 50 | 7576 | ||
XM_005251773.2 | XP_005251830.1 | 50 | 7491 | |||
XM_005251776.2 | XP_005251833.1 | 49 | 7331 | |||
XM_011518339.2 | XP_011516641.1 | 51 | 7566 | |||
XM_011518340.2 | XP_011516642.1 | 52 | 7781 | |||
XM_011518341.2 | XP_011516643.1 | 51 | 7560 | |||
XM_011518342.2 | XP_011516644.1 | 48 | 7269 | |||
XM_017014378.1 | XP_016869867.1 | 51 | 7764 | |||
XM_017014379.1 | XP_016869868.1 | 51 | 8172 | |||
XM_017014380.1 | XP_016869869.1 | 51 | 7498 | |||
XM_017014381.1 | XP_016869870.1 | 51 | 7564 | |||
XM_017014382.1 | XP_016869871.1 | 49 | 8029 | |||
GenBank mRNA | AB208839.1 | - | 11 | 4211 | ||
AF165281.1 | - | 48 | 7111 | |||
AF285167.1 | - | 50 | 7461 | |||
AK027864.1 | - | 15 | 2293 | |||
BC146856.1 | - | 50 | 7385 | 9056784 |