Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005630.2 | NP_005621.2 | 6 | 1078 | ||
XM_017007075.1 | XP_016862564.1 | 6 | 1021 | |||
XM_017007076.1 | XP_016862565.1 | 6 | 1021 | |||
XM_017007077.1 | XP_016862566.1 | 7 | 1066 | |||
GenBank mRNA | AK309561.1 | - | 7 | 1066 | ||
BC041140.1 | - | 6 | 907 | 5751516 | ||
BC051347.1 | - | 6 | 987 | 6481104 | ||
U70867.1 | - | 6 | 887 |