Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005692.4 | NP_005683.2 | 4 | 734 | ||
NM_007189.2 | NP_009120.1 | 4 | 734 | |||
XM_017011639.1 | XP_016867128.1 | 4 | 611 | |||
XM_017011640.1 | XP_016867129.1 | 4 | 696 | |||
GenBank mRNA | AF261091.1 | - | 4 | 591 | ||
AF261092.1 | - | 4 | 572 | |||
AJ005016.1 | - | 2 | 261 | |||
AL050291.1 | - | 3 | 526 | |||
BC001661.2 | - | 4 | 566 | 2988125 | ||
BT007451.1 | - | 3 | 480 | |||
CR987440.1 | - | 3 | 465 |