Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005692.4 | NP_005683.2 | 8 | 1255 | ||
NM_007189.2 | NP_009120.1 | 8 | 1255 | |||
XM_017011639.1 | XP_016867128.1 | 8 | 1132 | |||
XM_017011640.1 | XP_016867129.1 | 8 | 1217 | |||
GenBank mRNA | AF261091.1 | - | 8 | 1112 | ||
AF261092.1 | - | 8 | 1093 | |||
AJ005016.1 | - | 6 | 782 | |||
AL050291.1 | - | 7 | 1047 | |||
BC001661.2 | - | 8 | 1087 | 2988125 | ||
BG720644.1 | - | 3 | 393 | |||
BT007451.1 | - | 7 | 1001 |