Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNUPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042581.1 | NP_001036046.1 | 3 | 510 | ||
NM_001042588.1 | NP_001036053.1 | 3 | 315 | |||
NM_005701.3 | NP_005692.1 | 3 | 615 | |||
GenBank mRNA | AF039029.1 | - | 3 | 260 | ||
AK054641.1 | - | 4 | 748 | |||
AK289475.1 | - | 3 | 311 | |||
BC004203.2 | - | 3 | 246 | 3605328 | ||
BU945130.1 | - | 3 | 409 | |||
CD244425.1 | - | 3 | 510 | |||
CR456811.1 | - | 2 | 170 |