Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297644.1 | NP_001284573.1 | 12 | 1368 | ||
NM_005869.3 | NP_005860.2 | 12 | 1368 | |||
GenBank mRNA | AF039692.1 | - | 12 | 1320 | ||
AF039693.1 | - | 8 | 759 | |||
AK225105.1 | - | 12 | 1335 | |||
AK308223.1 | - | 12 | 1338 | |||
AK312954.1 | - | 12 | 1338 | |||
AY358569.1 | - | 12 | 1329 | |||
BC012117.1 | - | 12 | 1321 | 4552442 |