Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006151.2 | NP_006142.1 | 3 | 450 | ||
XM_011524808.2 | XP_011523110.1 | 3 | 326 | |||
GenBank mRNA | AK131496.1 | - | 2 | 1113 | ||
AK308378.1 | - | 3 | 175 | |||
AK310965.1 | - | 3 | 205 | |||
BC107166.1 | - | 3 | 321 | 40034365 | ||
BC107167.1 | - | 3 | 321 | 40034371 | ||
BX487327.1 | - | 3 | 389 | |||
BX648309.1 | - | 3 | 288 | |||
DB230339.1 | - | 3 | 450 | |||
U39573.1 | - | 3 | 215 |