Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006224.3 | NP_006215.1 | 7 | 676 | ||
XM_011523939.1 | XP_011522241.1 | 6 | 588 | |||
XM_011523940.1 | XP_011522242.1 | 7 | 1098 | |||
GenBank mRNA | AK303986.1 | - | 7 | 446 | ||
AL542612.3 | - | 7 | 674 | |||
BC027177.1 | - | 5 | 523 | |||
BC045108.1 | - | 7 | 651 | 5454273 | ||
BC068538.1 | - | 7 | 559 | |||
BC082976.1 | - | 7 | 591 | 6578454 | ||
D30036.1 | - | 7 | 628 | |||
EU794647.1 | - | 7 | 651 | |||
M73704.1 | - | 7 | 636 |