Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098486.1 | NP_001091956.1 | 11 | 1412 | ||
NM_006632.3 | NP_006623.2 | 10 | 1178 | |||
GenBank mRNA | AK298271.1 | - | 11 | 1412 | ||
BC017952.1 | - | 10 | 1147 | 4557795 | ||
BX107915.1 | - | 1 | 24 | |||
CB163247.1 | - | 5 | 550 | |||
U90545.1 | - | 10 | 1387 |