Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207055.1 | NP_001193984.1 | 16 | 2099 | ||
NM_001207056.1 | NP_001193985.1 | 15 | 1997 | |||
NM_006892.3 | NP_008823.1 | 18 | 2285 | |||
NM_175848.1 | NP_787044.1 | 17 | 2225 | |||
NM_175849.1 | NP_787045.1 | 17 | 2225 | |||
NM_175850.2 | NP_787046.1 | 17 | 2204 | |||
XM_011528653.2 | XP_011526955.1 | 17 | 2077 | |||
XM_011528654.2 | XP_011526956.1 | 16 | 1952 | |||
GenBank mRNA | AB208880.1 | - | 17 | 2449 | ||
AF129267.1 | - | 1 | 59 | |||
AF129268.1 | - | 1 | 59 | |||
AF129269.1 | - | 1 | 59 | |||
AF156487.1 | - | 17 | 2018 | |||
AF156488.1 | - | 18 | 2078 | |||
AF176228.1 | - | 17 | 2204 | |||
AF331857.1 | - | 18 | 2285 | |||
AK001191.1 | - | 11 | 1157 | |||
AK295176.1 | - | 15 | 1699 | |||
AK299915.1 | - | 16 | 1876 | |||
DQ321787.1 | - | 17 | 2319 |