Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288985.1 | NP_001275914.1 | 5, 6 | 814 | ||
NM_001288986.1 | NP_001275915.1 | 4, 5 | 653 | |||
NM_007168.3 | NP_009099.1 | 4, 5 | 653 | |||
XM_005256938.2 | XP_005256995.1 | 4, 5 | 681 | |||
XM_005256940.4 | XP_005256997.1 | 3, 4 | 586 | |||
XM_011524191.2 | XP_011522493.1 | 4, 5 | 644 | |||
XM_011524192.2 | XP_011522494.1 | 4, 5 | 644 | |||
GenBank mRNA | AB020629.1 | - | 4, 5 | 612 | ||
AK295876.1 | - | 4, 5 | 626 | |||
AK304286.1 | - | 3, 4 | 485 | |||
AK304360.1 | - | 4, 5 | 624 | |||
BC024003.1 | - | 4, 5 | 658 | |||
BC037309.1 | - | 4, 5 | 649 | |||
BC047765.1 | - | 5, 6 | 773 | |||
BC094689.1 | - | 4, 5 | 612 | |||
BC130280.1 | - | 5, 6 | 663 | 40146311 |