Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007171.3 | NP_009102.3 | 8 | 894 | ||
XM_005272156.1 | XP_005272213.1 | 8 | 1020 | |||
XM_005272158.1 | XP_005272215.1 | 7 | 749 | |||
XM_005272159.1 | XP_005272216.1 | 6 | 639 | |||
XM_005272162.2 | XP_005272219.1 | 8 | 868 | |||
XM_006716932.1 | XP_006716995.1 | 7 | 672 | |||
XM_011518140.1 | XP_011516442.1 | 7 | 747 | |||
XM_011518142.1 | XP_011516444.1 | 6 | 549 | |||
XM_011518145.1 | XP_011516447.1 | 5 | 415 | |||
XM_017014203.1 | XP_016869692.1 | 8 | 1342 | |||
XM_017014205.1 | XP_016869694.1 | 8 | 816 | |||
GenBank mRNA | AB209430.1 | - | 4 | 1754 | ||
AF095136.1 | - | 8 | 871 | |||
AK301486.1 | - | 7 | 764 | |||
AK303254.1 | - | 5 | 547 | |||
AK307876.1 | - | 8 | 853 | |||
AK315218.1 | - | 8 | 829 |