Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002567.3 | NP_002558.1 | 2 | 561 | ||
GenBank mRNA | AK226006.1 | - | 2 | 340 | ||
AK299414.1 | - | 2 | 320 | |||
AK308056.1 | - | 2 | 319 | |||
AK311927.1 | - | 2 | 332 | |||
BC008714.2 | - | 2 | 334 | 2822606 | ||
BC017396.1 | - | 2 | 355 | 4074533 | ||
BC018799.1 | - | 2 | 325 | |||
BC031102.1 | - | 2 | 323 | 5300366 | ||
BF978793.1 | - | 2 | 403 | |||
D16111.1 | - | 2 | 326 | |||
EU668351.1 | - | 1 | 94 | |||
EU794643.1 | - | 2 | 325 | |||
GQ472218.1 | - | 1 | 94 | |||
HM131449.1 | - | 2 | 317 | |||
S76773.1 | - | 1 | 94 | |||
X75252.1 | - | 2 | 345 | |||
X85033.1 | - | 2 | 238 |