Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012089.2 | NP_036221.2 | 4, 5 | 1107 | ||
XM_011544135.2 | XP_011542437.1 | 4, 5 | 1878 | |||
XM_011544136.2 | XP_011542438.1 | 2, 3 | 344 | |||
GenBank mRNA | AB013380.1 | - | 4, 5 | 1097 | ||
AF216833.1 | - | 4, 5 | 1107 | |||
AK172794.1 | - | 3, 4 | 363 | |||
BC064930.1 | - | 4, 5 | 1102 | 6143235 |