Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012434.4 | NP_036566.1 | 4, 5 | 752 | ||
XM_005248710.3 | XP_005248767.1 | 4, 5 | 774 | |||
XM_005248711.2 | XP_005248768.1 | 4, 5 | 1006 | |||
XM_011535750.2 | XP_011534052.1 | 4, 5 | 710 | |||
XM_017010730.1 | XP_016866219.1 | 3, 4 | 918 | |||
XM_017010731.1 | XP_016866220.1 | 3, 4 | 768 | |||
GenBank mRNA | AF244577.1 | - | 5, 6 | 1009 | ||
AJ387747.1 | - | 4, 5 | 894 | |||
AK075320.1 | - | 4, 5 | 706 | |||
AK294068.1 | - | 3, 4 | 623 | |||
AK315789.1 | - | 4, 5 | 710 | |||
BC020961.2 | - | 4, 5 | 711 | 3847279 |