Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318192.1 | NP_001305121.1 | 16 | 2860 | ||
NM_012450.3 | NP_036582.2 | 16 | 2857 | |||
XM_011516024.2 | XP_011514326.1 | 15 | 2502 | |||
XM_017011962.1 | XP_016867451.1 | 14 | 2228 | |||
GenBank mRNA | AB208809.1 | - | 13 | 7135 | ||
BC030689.2 | - | 16 | 2839 | 4821193 |