Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145044.1 | NP_001138516.1 | 2 | 721 | ||
NM_013272.3 | NP_037404.2 | 2 | 721 | |||
NR_135775.1 | 2 | 451 | ||||
XM_005254889.1 | XP_005254946.1 | 2 | 610 | |||
XM_005254891.2 | XP_005254948.1 | 2 | 440 | |||
XM_011521456.2 | XP_011519758.1 | 3 | 521 | |||
GenBank mRNA | AB031050.2 | - | 2 | 713 | ||
AF187816.1 | - | 2 | 520 | |||
AF205074.1 | - | 2 | 709 | |||
AK097797.1 | - | 3 | 521 | |||
AK290872.1 | - | 2 | 559 | |||
AK310306.1 | - | 2 | 416 | |||
BC000585.2 | - | 2 | 573 | 3163826 | ||
HY004505.1 | - | 2 | 454 |