Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014228.4 | NP_055043.2 | 14 | 3485 | ||
XM_017009767.1 | XP_016865256.1 | 15 | 3392 | |||
XM_017009768.1 | XP_016865257.1 | 15 | 3468 | |||
XM_017009769.1 | XP_016865258.1 | 15 | 4999 | |||
XM_017009770.1 | XP_016865259.1 | 15 | 3119 | |||
GenBank mRNA | AK096607.1 | - | 14 | 3389 |