Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014485.2 | NP_055300.1 | 3 | 285 | ||
GenBank mRNA | AK290075.1 | - | 3 | 285 | ||
BC020734.1 | - | 3 | 262 | 4731411 | ||
CB988786.1 | - | 312 | ||||
CR541662.1 | - | 2 | 194 | |||
CR541679.1 | - | 2 | 194 | |||
D82073.1 | - | 2 | 204 |