Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015696.4 | NP_056511.2 | 3 | 897 | ||
GenBank mRNA | AF091092.1 | - | 2 | 638 | ||
AF320068.2 | - | 3 | 896 | |||
AK027683.1 | - | 3 | 897 | |||
AY358402.1 | - | 3 | 891 | |||
BC032788.1 | - | 3 | 870 | 5212085 |