Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015913.3 | NP_056997.1 | 3, 4 | 1294 | ||
GenBank mRNA | AF131758.1 | - | 3, 4 | 329 | ||
AF543416.1 | - | 3, 4 | 498 | |||
AK075409.1 | - | 3, 4 | 292 | |||
AY358982.1 | - | 3, 4 | 325 | |||
BC001493.2 | - | 3, 4 | 295 | 3506109 | ||
BC008913.2 | - | 3, 4 | 324 | 3659407 | ||
BC008953.2 | - | 3, 4 | 273 | 2959502 |