Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015913.3 | NP_056997.1 | 7 | 2286 | ||
GenBank mRNA | AF131758.1 | - | 7 | 1321 | ||
AF543416.1 | - | 7 | 1490 | |||
AK056169.1 | - | 7 | 2286 | |||
AK075409.1 | - | 7 | 1284 | |||
BC001493.2 | - | 7 | 1287 | 3506109 | ||
BC008913.2 | - | 7 | 1316 | 3659407 | ||
BC008953.2 | - | 7 | 1266 | 2959502 |