Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145099.1 | NP_001138571.1 | 9 | 1811 | ||
NM_017585.3 | NP_060055.2 | 10 | 1997 | |||
XM_011518189.2 | XP_011516491.2 | 10 | 1974 | |||
XM_017014237.1 | XP_016869726.1 | 10 | 1965 | |||
XM_017014238.1 | XP_016869727.1 | 10 | 1969 | |||
GenBank mRNA | AB209108.1 | - | 4 | 5230 | ||
AJ011372.2 | - | 10 | 1939 | |||
AK074836.1 | - | 9 | 1761 | |||
AK074927.1 | - | 10 | 1980 | |||
AK096630.1 | - | 8 | 2874 | |||
AK222919.1 | - | 10 | 1942 | |||
AK225106.1 | - | 10 | 1870 | |||
BC013740.1 | - | 10 | 1931 | 4449594 |