Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017946.3 | NP_060416.1 | 2 | 568 | ||
NR_046478.1 | 2 | 568 | ||||
GenBank mRNA | AK000738.1 | - | 2 | 420 | ||
AY358643.1 | - | 2 | 448 | |||
BC005206.1 | - | 2 | 451 | 4042173 | ||
DB498479.1 | - | 2 | 188 | |||
JQ797475.1 | - | 2 | 275 |