Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018420.2 | NP_060890.2 | 3 | 531 | ||
XM_005271004.3 | XP_005271061.1 | 3 | 492 | |||
GenBank mRNA | AK125072.1 | - | 1 | 100 | ||
AY145501.1 | - | 3 | 401 | |||
AY358258.1 | - | 3 | 415 | |||
BC026358.1 | - | 3 | 530 | 4814675 | ||
KU178698.1 | - | 3 | 365 | |||
KU178699.1 | - | 3 | 365 | |||
KU178700.1 | - | 3 | 365 | |||
KU178701.1 | - | 2 | 152 | |||
KU178702.1 | - | 3 | 365 | |||
KU178703.1 | - | 2 | 152 |