Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018420.2 | NP_060890.2 | 7 | 1094 | ||
GenBank mRNA | AK125072.1 | - | 5 | 662 | ||
AY145501.1 | - | 7 | 964 | |||
BC026358.1 | - | 7 | 1093 | 4814675 | ||
KU178698.1 | - | 7 | 928 | |||
KU178699.1 | - | 8 | 1259 | |||
KU178700.1 | - | 7 | 928 | |||
KU178701.1 | - | 9 | 1046 | |||
KU178702.1 | - | 7 | 851 | |||
KU178703.1 | - | 6 | 715 |