Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320537.1 | NP_001307466.1 | 20 | 3256 | ||
NM_018964.3 | NP_061837.3 | 21 | 2625 | |||
XM_011529615.1 | XP_011527917.1 | 19 | 3397 | |||
XM_017028376.1 | XP_016883865.1 | 18 | 3332 | |||
XM_017028377.1 | XP_016883866.1 | 21 | 2550 | |||
XM_017028378.1 | XP_016883867.1 | 21 | 3009 | |||
XM_017028379.1 | XP_016883868.1 | 22 | 2656 | |||
XM_017028380.1 | XP_016883869.1 | 20 | 2456 | |||
XM_017028381.1 | XP_016883870.1 | 20 | 2485 | |||
XM_017028382.1 | XP_016883871.1 | 19 | 3738 | |||
GenBank mRNA | AF289589.1 | - | 1 | 1909 | ||
AF311320.1 | - | 20 | 3195 | |||
AJ269529.2 | - | 21 | 2625 | |||
AK025993.1 | - | 7 | 1088 | |||
AL355683.1 | - | 1 | 620 |